Search
Search

OASA1| Gene Symbol: OASA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203803.1 | NP_001190732.1 | 7 - 8 | 944 | 132 | |
| NM_001203804.1 | NP_001190733.1 | 7 - 8 | 947 | 132 | ||
| NM_001340975.1 | NP_001328013.1 | 7 - 8 | 1126 | 132 | ||
| NM_117574.4 | NP_193224.1 | 7 - 8 | 931 | 132 | ||
| NM_179055.3 | NP_849386.1 | 7 - 8 | 801 | 132 | ||
| GenBank mRNA | BT025878.1 | - | 6 - 7 | 653 | 132 | |
| X80376.2 | - | 714 | 132 | |||
Set Membership: |
|