Search
Search

NAC071| Gene Symbol: NAC071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341249.1 | NP_001329603.1 | 1 - 2 | 677 | 154 | |
| NM_117908.2 | NP_193532.1 | 1 - 2 | 455 | 154 | ||
| GenBank mRNA | LC168677.1 | - | 1 - 2 | 160 | 154 | |
Set Membership: |
|