Search
Search

AT1G70650| Gene Symbol: AT1G70650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198435.1 | NP_001185364.1 | 4 - 5 | 941 | 79 | |
| NM_105733.3 | NP_564993.1 | 4 - 5 | 940 | 79 | ||
Set Membership: |
|