Search
Search

HDG11| Gene Symbol: HDG11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105996.4 | NP_177479.1 | 5 - 6 | 1899 | 88 | |
| GenBank mRNA | BT003979.1 | - | 5 - 6 | 1640 | 88 | |
| BT004915.1 | - | 5 - 6 | 1034 | 88 | ||
Set Membership: |
|