Search
Search

TIF3H1| Gene Symbol: TIF3H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100960.3 | NP_563880.1 | 4 - 5 | 628 | 76 | |
| NM_202079.1 | NP_973808.1 | 3 - 4 | 514 | 76 | ||
| GenBank mRNA | AY054641.1 | - | 4 - 5 | 462 | 76 | |
| AY081546.1 | - | 3 - 4 | 311 | 76 | ||
| AY087338.1 | - | 4 - 5 | 530 | 76 | ||
| BT000765.1 | - | 4 - 5 | 407 | 76 | ||
Set Membership: |
|