Search
Search

SAT32| Gene Symbol: SAT32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102541.3 | NP_564294.1 | 10 - 11 | 1493 | 81 | |
| NM_179384.2 | NP_849715.2 | 9 - 10 | 1668 | 81 | ||
| NM_202194.3 | NP_973923.1 | 10 - 11 | 1581 | 81 | ||
| GenBank mRNA | AF361609.1 | - | 10 - 11 | 1478 | 81 | |
| AF428281.1 | - | 10 - 11 | 1478 | 81 | ||
| AY078052.1 | - | 9 - 10 | 1182 | 81 | ||
Set Membership: |
|