Search
Search

BGLU41| Gene Symbol: BGLU41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345089.1 | NP_001330205.1 | 5 - 6 | 821 | 69 | |
| NM_001345090.1 | NP_001330204.1 | 8 - 9 | 1112 | 69 | ||
| NM_124837.3 | NP_200268.3 | 8 - 9 | 1112 | 69 | ||
Set Membership: |
|