Search
Search

EX1| Gene Symbol: EX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342225.1 | NP_001328575.1 | 2 - 3 | 629 | 74 | |
| NM_119519.4 | NP_567929.1 | 2 - 3 | 779 | 74 | ||
| NM_179157.3 | NP_849488.1 | 2 - 3 | 779 | 74 | ||
| GenBank mRNA | AY059742.1 | - | 2 - 3 | 568 | 74 | |
| AY117173.1 | - | 2 - 3 | 511 | 74 | ||
Set Membership: |
|