Search
Search

HCF153| Gene Symbol: HCF153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119305.4 | NP_194884.1 | 1 - 2 | 364 | 63 | |
| GenBank mRNA | AF324683.2 | - | 1 - 2 | 297 | 63 | |
| AF326897.1 | - | 1 - 2 | 297 | 63 | ||
| AF339716.1 | - | 1 - 2 | 283 | 63 | ||
| AF372916.1 | - | 1 - 2 | 297 | 63 | ||
| AY086252.1 | - | 1 - 2 | 301 | 63 | ||
| BT002676.1 | - | 1 - 2 | 283 | 63 | ||
Set Membership: |
|