Search
Search

AT3G15680| Gene Symbol: AT3G15680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338181.1 | NP_001326186.1 | 3 - 4 | 1056 | 68 | |
| NM_112438.4 | NP_188189.1 | 3 - 4 | 842 | 68 | ||
Set Membership: |
|