Search
Search

DELTA-TIP| Gene Symbol: DELTA-TIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112495.4 | NP_188245.1 | 1 - 2 | 298 | 106 | |
| GenBank mRNA | AK229954.1 | - | 1670 | 106 | ||
| AY085921.1 | - | 1 - 2 | 196 | 106 | ||
| U39485.1 | - | 1 - 2 | 141 | 106 | ||
Set Membership: |
|