Search
Search

XPL1| Gene Symbol: XPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112681.3 | NP_188427.2 | 4 - 5 | 626 | 64 | |
| GenBank mRNA | AF197940.1 | - | 4 - 5 | 612 | 64 | |
| AF367299.1 | - | 4 - 5 | 551 | 64 | ||
| AY058175.1 | - | 4 - 5 | 626 | 64 | ||
| AY091683.1 | - | 4 - 5 | 411 | 64 | ||
Set Membership: |
|