Search
Search

AGF1| Gene Symbol: AGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_119705.3 | NP_195265.2 | 1 | 1045 | 118 | |
| GenBank mRNA | BT014971.1 | - | 1 | 1001 | 118 | |
| BT015845.1 | - | 1 | 768 | 118 | ||
Set Membership: |
|