Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRP39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160830.2 | NP_001154302.1 | 9 - 10 | 2052 | 64 | |
NM_001197976.1 | NP_001184905.1 | 14 - 15 | 2571 | 64 | ||
NM_001331460.1 | NP_001322270.1 | 11 - 12 | 2461 | 64 | ||
NM_001331461.1 | NP_001322271.1 | 9 - 10 | 1872 | 64 | ||
NM_100289.4 | NP_563700.1 | 12 - 13 | 2375 | 64 | ||
GenBank mRNA | AY056321.1 | - | 12 - 13 | 2340 | 64 | |
AY087878.1 | - | 12 - 13 | 2342 | 64 | ||
AY091187.1 | - | 11 - 12 | 2192 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |