Search
Search

AT1G04850| Gene Symbol: AT1G04850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331503.1 | NP_001323139.1 | 8 - 9 | 994 | 68 | |
| NM_100363.5 | NP_563718.1 | 8 - 9 | 1009 | 68 | ||
| GenBank mRNA | AY062609.1 | - | 8 - 9 | 808 | 68 | |
| AY088435.1 | - | 8 - 9 | 862 | 68 | ||
| BT000147.1 | - | 8 - 9 | 775 | 68 | ||
Set Membership: |
|