Search
Search

MYB60| Gene Symbol: MYB60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100755.3 | NP_172358.1 | 2 - 3 | 410 | 117 | |
| GenBank mRNA | AK117469.1 | - | 2 - 3 | 280 | 117 | |
| AY519551.1 | - | 2 - 3 | 261 | 117 | ||
| BT005074.1 | - | 2 - 3 | 261 | 117 | ||
Set Membership: |
|