Search
Search

RAB11c| Gene Symbol: RAB11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100835.3 | NP_172434.1 | 1 - 2 | 409 | 67 | |
| GenBank mRNA | AY045620.1 | - | 1 - 2 | 212 | 67 | |
| AY085684.1 | - | 1 - 2 | 207 | 67 | ||
| AY133657.1 | - | 1 - 2 | 165 | 67 | ||
| U74669.1 | - | 1 - 2 | 265 | 67 | ||
Set Membership: |
|