Search
Search

GS1| Gene Symbol: GS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125126.3 | NP_568858.1 | 5 - 6 | 713 | 64 | |
| GenBank mRNA | AY063967.1 | - | 5 - 6 | 621 | 64 | |
| AY084531.1 | - | 5 - 6 | 621 | 64 | ||
| AY114031.1 | - | 5 - 6 | 592 | 64 | ||
Set Membership: |
|