Search
Search

ORP3C| Gene Symbol: ORP3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_125333.5 | NP_200750.1 | 5 - 6 | 663 | 74 | |
| GenBank mRNA | AY054473.1 | - | 5 - 6 | 616 | 74 | |
| BT000115.1 | - | 5 - 6 | 499 | 74 | ||
Set Membership: |
|