Search
Search

CLPR1| Gene Symbol: CLPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103884.4 | NP_564560.1 | 1 - 2 | 428 | 140 | |
| GenBank mRNA | AB022330.1 | - | 1 - 2 | 383 | 140 | |
| AY045677.1 | - | 1 - 2 | 340 | 140 | ||
| BT000572.1 | - | 1 - 2 | 236 | 140 | ||
Set Membership: |
|