Search
Search

VAMP713| Gene Symbol: VAMP713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343158.1 | NP_001331552.1 | 3 - 4 | 840 | 88 | |
| NM_121153.5 | NP_196676.1 | 3 - 4 | 846 | 88 | ||
| GenBank mRNA | AY091002.1 | - | 3 - 4 | 749 | 88 | |
| AY122934.1 | - | 3 - 4 | 573 | 88 | ||
Set Membership: |
|