Search
Search

AT1G12650| Gene Symbol: AT1G12650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035954.4 | NP_001031031.1 | 3 - 4 | 485 | 64 | |
| NM_001084055.3 | NP_001077524.1 | 3 - 4 | 416 | 64 | ||
| NM_001084056.1 | NP_001077525.1 | 3 - 4 | 322 | 64 | ||
| NM_101135.3 | NP_172725.2 | 2 - 3 | 357 | 64 | ||
| GenBank mRNA | AF446361.1 | - | 2 - 3 | 357 | 64 | |
| AY101527.1 | - | 2 - 3 | 233 | 64 | ||
Set Membership: |
|