Search
Search

RL6| Gene Symbol: RL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084356.3 | NP_001077825.1 | 1 | 344 | 102 | |
| NM_106181.2 | NP_177661.1 | 1 | 216 | 102 | ||
| GenBank mRNA | AY519525.1 | - | 1 | 216 | 102 | |
| BT026091.1 | - | 1 | 216 | 102 | ||
| DQ395346.1 | - | 1 | 216 | 102 | ||
Set Membership: |
|