Search
Search

AT1G77460| Gene Symbol: AT1G77460 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198490.2 | NP_001185419.1 | 4 - 5 | 3411 | 100 | |
| NM_001334782.1 | NP_001319395.1 | 4 - 5 | 3336 | 100 | ||
| NM_001334783.1 | NP_001322685.1 | 4 - 5 | 3388 | 100 | ||
| NM_106395.4 | NP_177870.2 | 3 - 4 | 3615 | 100 | ||
Set Membership: |
|