Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036229.3 | NP_001031306.2 | 5 - 6 | 689 | 100 | |
NM_001124151.1 | NP_001117623.1 | 5 - 6 | 680 | 100 | ||
NM_001198516.1 | NP_001185445.1 | 5 - 6 | 672 | 100 | ||
NM_106643.5 | NP_178112.2 | 5 - 6 | 661 | 100 | ||
GenBank mRNA | AK227714.1 | - | 5 - 6 | 607 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |