Search
Search

APT2| Gene Symbol: APT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334951.1 | NP_001320554.1 | 4 - 5 | 808 | 70 | |
| NM_106654.3 | NP_178122.1 | 5 - 6 | 1250 | 70 | ||
| GenBank mRNA | AY072021.1 | - | 5 - 6 | 564 | 70 | |
| BT001122.1 | - | 5 - 6 | 501 | 70 | ||
| X96866.1 | - | 523 | 70 | |||
Set Membership: |
|