Search
Search

GSTU8| Gene Symbol: GSTU8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111761.3 | NP_187538.1 | 1 - 2 | 787 | 66 | |
| GenBank mRNA | AY086116.1 | - | 1 - 2 | 388 | 66 | |
| BT024844.1 | - | 1 - 2 | 327 | 66 | ||
Set Membership: |
|