Search
Search

HKT1| Gene Symbol: HKT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117099.6 | NP_567354.1 | 1 - 2 | 1225 | 74 | |
| GenBank mRNA | AK228564.1 | - | 1 - 2 | 1095 | 74 | |
| BT005751.1 | - | 1 - 2 | 1095 | 74 | ||
Set Membership: |
|