Search
Search

ABCE2| Gene Symbol: ABCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118041.5 | NP_193656.2 | 4 - 5 | 497 | 130 | |
| GenBank mRNA | AK226939.1 | - | 4 - 5 | 404 | 130 | |
| AY099697.1 | - | 4 - 5 | 417 | 130 | ||
| BT000298.1 | - | 3 - 4 | 293 | 130 | ||
Set Membership: |
|