Search
Search

LCB2| Gene Symbol: LCB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036855.1 | NP_001031932.1 | 2 - 3 | 422 | 93 | |
| NM_122272.3 | NP_197756.1 | 2 - 3 | 438 | 93 | ||
| GenBank mRNA | AY050829.1 | - | 2 - 3 | 385 | 93 | |
| AY133827.1 | - | 2 - 3 | 190 | 93 | ||
Set Membership: |
|