Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: COX6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102094.4 | NP_173661.1 | 3 - 4 | 765 | 70 | |
GenBank mRNA | AB035444.1 | - | 3 - 4 | 467 | 70 | |
AY063997.1 | - | 3 - 4 | 572 | 70 | ||
AY096696.1 | - | 3 - 4 | 439 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |