Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ERD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332846.1 | NP_001321392.1 | 5 - 6 | 770 | 74 | |
NM_102673.6 | NP_564326.1 | 5 - 6 | 940 | 74 | ||
GenBank mRNA | AY093240.1 | - | 5 - 6 | 748 | 74 | |
BT009655.1 | - | 5 - 6 | 614 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |