Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RING1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344539.1 | NP_001331587.1 | 2 - 3 | 134 | 112 | |
NM_123795.5 | NP_199241.2 | 3 - 4 | 608 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |