Search
Search

AT5G47930| Gene Symbol: AT5G47930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124167.4 | NP_199604.1 | 2 - 3 | 219 | 70 | |
| GenBank mRNA | AY046032.1 | - | 2 - 3 | 168 | 70 | |
| AY079419.1 | - | 1 - 2 | 99 | 70 | ||
Set Membership: |
|