Search
Search

HVA22E| Gene Symbol: HVA22E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124450.5 | NP_568744.1 | 2 - 3 | 185 | 88 | |
| GenBank mRNA | AF290892.1 | - | 2 - 3 | 107 | 88 | |
| AY084933.1 | - | 2 - 3 | 119 | 88 | ||
| BT028898.1 | - | 2 - 3 | 69 | 88 | ||
Set Membership: |
|