Search
Search

AGP22| Gene Symbol: AGP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124704.3 | NP_200137.1 | 1 - 2 | 265 | 95 | |
| GenBank mRNA | AK228915.1 | - | 1 - 2 | 214 | 95 | |
| BT010777.1 | - | 1 - 2 | 184 | 95 | ||
| BT011258.1 | - | 1 - 2 | 119 | 95 | ||
Set Membership: |
|