Search
Search

AT2G37600| Gene Symbol: AT2G37600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084548.1 | NP_001078017.1 | 2 - 3 | 372 | 153 | |
| NM_129316.4 | NP_181296.1 | 3 - 4 | 367 | 153 | ||
| GenBank mRNA | AY052221.1 | - | 3 - 4 | 330 | 153 | |
| AY086261.1 | - | 3 - 4 | 335 | 153 | ||
Set Membership: |
|