Search
Search

MPK1| Gene Symbol: MPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035940.2 | NP_001031017.1 | 1 - 2 | 783 | 122 | |
| NM_001331892.1 | NP_001320530.1 | 2 - 3 | 538 | 122 | ||
| NM_100895.3 | NP_172492.1 | 2 - 3 | 675 | 122 | ||
| GenBank mRNA | AY059937.1 | - | 2 - 3 | 504 | 122 | |
| BT000062.1 | - | 1 - 2 | 424 | 122 | ||
Set Membership: |
|