Search
Search

AT3G01850| Gene Symbol: AT3G01850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337361.1 | NP_001326139.1 | 5 - 6 | 409 | 141 | |
| NM_001337362.1 | NP_001326138.1 | 5 - 6 | 409 | 141 | ||
| NM_111051.3 | NP_566153.1 | 6 - 7 | 772 | 141 | ||
| NM_180164.4 | NP_850495.1 | 6 - 7 | 892 | 141 | ||
| GenBank mRNA | AF370479.1 | - | 6 - 7 | 532 | 141 | |
| AY086329.1 | - | 6 - 7 | 513 | 141 | ||
| BT002174.1 | - | 6 - 7 | 482 | 141 | ||
Set Membership: |
|