Search
Search

LHCB2.3| Gene Symbol: LHCB2.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338904.1 | NP_001325794.1 | 1 - 2 | 246 | 173 | |
| NM_113685.4 | NP_189406.1 | 1 - 2 | 211 | 173 | ||
| GenBank mRNA | AF134125.1 | - | 1 - 2 | 155 | 173 | |
| AF370557.1 | - | 1 - 2 | 199 | 173 | ||
| BT006298.1 | - | 1 - 2 | 150 | 173 | ||
Set Membership: |
|