Search
Search

MSRB7| Gene Symbol: MSRB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084953.1 | NP_001078422.1 | 4 | 527 | 159 | |
| NM_118303.4 | NP_567637.1 | 3 | 779 | 159 | ||
| GenBank mRNA | AY072355.1 | - | 3 | 628 | 159 | |
Set Membership: |
|