Search
Search

SCRL16| Gene Symbol: SCRL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036265.2 | NP_001031342.1 | 1 - 2 | 130 | 84 | |
| NM_001335310.1 | NP_001325116.1 | 1 - 2 | 130 | 84 | ||
Set Membership: |
|