Search
Search

AT1G68200| Gene Symbol: AT1G68200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084323.1 | NP_001077792.1 | 1 - 2 | 592 | 105 | |
| NM_001334355.1 | NP_001319345.1 | 2 - 3 | 695 | 105 | ||
| GenBank mRNA | DQ446410.1 | - | 1 - 2 | 592 | 105 | |
Set Membership: |
|