Search
Search

IFNT2
IFNT
IFNT3
LOC112445660| Gene Symbol: IFNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015511.4 | NP_001015511.3 | 1 | 967 | 145 | |
| GenBank mRNA | M31557.2 | - | 1 | 753 | 145 | |
| Gene Symbol: IFNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168279.1 | NP_001161751.1 | 1 | 145 | ||
| GenBank mRNA | CV982415.1 | - | 1 | 145 | ||
| Gene Symbol: IFNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168275.1 | NP_001161747.1 | 1 | 145 | ||
| Gene Symbol: LOC112445660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M31558.1 | - | 1 - 2 | 140 | ||
Set Membership: |
|