Search
Search

ISG15| Gene Symbol: ISG15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174366.1 | NP_776791.1 | 1 - 2 | 61 | 85 | |
| GenBank mRNA | BC102318.1 | - | 1 - 2 | 75 | 85 | |
| U96014.1 | - | 1 - 2 | 78 | 85 | ||
Set Membership: |
|