Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174654.2 | NP_777079.1 | 3 - 4 | 500 | 68 | |
GenBank mRNA | AJ508718.1 | - | 3 - 4 | 500 | 68 | |
AY052775.1 | - | 4 - 5 | 584 | 68 | ||
BC133437.1 | - | 3 - 4 | 545 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |