Search
Search

TMEM143| Gene Symbol: TMEM143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037447.2 | NP_001032524.1 | 3 - 4 | 365 | 77 | |
| GenBank mRNA | BC108117.1 | - | 3 - 4 | 366 | 77 | |
| BT029876.1 | - | 3 - 4 | 363 | 77 | ||
Set Membership: |
|