Search
Search

IVNS1ABP| Gene Symbol: IVNS1ABP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046184.4 | NP_001039649.2 | 3 - 4 | 378 | 134 | |
| GenBank mRNA | BC112450.1 | - | 5 - 6 | 639 | 134 | |
Set Membership: |
|