Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046412.2 | NP_001039877.1 | 4 - 5 | 635 | 122 | |
GenBank mRNA | BC105422.1 | - | 4 - 5 | 638 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |