Search
Search

SNAP23| Gene Symbol: SNAP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075831.1 | NP_001069299.1 | 4 - 5 | 252 | 74 | |
| GenBank mRNA | BC111350.1 | - | 4 - 5 | 252 | 74 | |
| BT030678.1 | - | 4 - 5 | 272 | 74 | ||
Set Membership: |
|